CAS RN 1185133-69-0
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
2-[(2-Azidoethoxy-d4)methyl]-4-(2-chlorophenyl)-3-ethoxycarbonyl-5-methoxycarbonyl)-6-methyl-1,4-dihydropyridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2-[(2-Azidoethoxy-d4)methyl]-4-(2-chlorophenyl)-3-ethoxycarbonyl-5-methoxycarbonyl)-6-methyl-1,4-dihydropyridine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.