CAS RN 1173-26-8
IUPAC Navn:
2-[(1S,3aS,3bS,9aR,9bS,10S,11aS)-10-hydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
Synonymer:
cort A
Molekylvægt (g/mol):
388.5
Molekylformel:
C23H32O5
InChi Key:
WKQCPUMQBMFPLC-ZWFCQKKLSA-N
SMILES:
CC(=O)OCC(=O)[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C
1
–
1
af
1
Resultater
1
Corticosterone 21-Acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.