CAS RN 1141892-37-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
4',6-Dihydroxy-3',5-dimethoxy-[1,1'-biphenyl]-3-carboxaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4',6-Dihydroxy-3',5-dimethoxy-[1,1'-biphenyl]-3-carboxaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.