CAS RN 1116-77-4
IUPAC Navn:
(4,4-diethoxybutyl)dimethylamine
Synonymer:
(4,4-diethoxybutyl)dimethylamine
Molekylvægt (g/mol):
189.3
Molekylformel:
C10H23NO2
InChi Key:
QKXMWBLNSPNBEY-UHFFFAOYSA-N
SMILES:
CCOC(CCCN(C)C)OCC
1
–
3
af
3
Resultater
1
4-Dimethylaminobutyraldehyde Diethyl Acetal, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-Dimethylaminobutyraldehyde Diethyl Acetal, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-Dimethylaminobutyraldehyde Diethyl Acetal, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.