Velkommen til Fisher Scientific
Summer Promotion
on Thermo Scientific Pierce Protein Research Products
NEW Thermo Scientific Pierce Protein Gels
Thermo Scientific Pierce Protein Gels use a special formulation to produce stronger, more resilient gels, making handling during and after electrophoresis easier. The extra stability of Pierce Protein Gels combined with the Tris-HEPES-SDS Running Buffer offers both speed and excellent resolution. Compatible Gel Tanks: Thermo Scientific Owl P82 System, Novex® XCell I, II™ and Surelock® Systems, C.B.S. Scientific CBDCX-700 Dual Cool System and PAGEr® Minigel Chamber.
NEW Thermo Scientific NanoDrop
Catalogue No Description Pkg. Size Pay Now!
pie-84708X2 Pierce Protein Gels, 4-8% SDS, 12-Well 20 gels 1.085,-
pie-84711X2 Pierce Protein Gels, 12% SDS, 12-Well 20 gels 1.085,-
pie-84713X2 Pierce Protein Gels, 4-20% SDS, 12-Well 20 gels 1.085,-
pie-84710X2 Pierce Protein Gels, 4-8% SDS, 17-Well 20 gels 1.085,-
pie-84712X2 Pierce Protein Gels, 12% SDS, 17-Well 20 gels 1.085,-
pie-84714X2 Pierce Protein Gels, 4-20% SDS, 17-Well 20 gels 1.085,-
Thermo Scientific Staining Kit and Reagents
Most assume that making coomassie gel stain saves money over using a premade stain. However, when you calculate the costs for the stain and destain reagents, Thermo Scientific Commassie-based Gel Stains are a more cost-effective option than making your own gel stain.
NEW Thermo Scientific NanoDrop
Catalogue No Description Pkg. Size Pay Now!
pie-24594 GelCode Blue Safe Protein Stain 1L 795,-
pie-24596 GelCode Blue Safe Protein Stain 3,5L 1.759,-
pie-24612 GelCode SilverSNAP Stain Kit II 1 Kit 1144,-
Thermo Scientific Pierce Fast Transfer System
The Thermo Scientific Pierce Fast Transfer System reduces protein transfer time from SDS-PAGE to a membrane from 45 minutes to only 10 minutes. The system consists of two parts, the Pierce Fast Semi-Dry Transfer Buffer and the Pierce Fast Semi-Dry Blotter. Use our cost-effective Transfer System and eliminate the need to invest in – and dispose of – consumables.
NEW Thermo Scientific NanoDrop
Catalogue No Description Pkg. Size Pay Now!
pie-88217 Pierce Fast Semi-Dry Blotter 1 Unit 9220,-
pie-35035 Pierce Fast Semi-Dry Transfer Buffer 500 ml 595,-
Thermo Scientific SuperSignal High Sensitivity Substrates
Chemiluminescence is the detection method of choice in most protein laboratories. It yields the greatest sensitivity of any available detection method and it allows multiple exposures to obtain the best image. The detection reagents can be removed and the entire blot reprobed to visualize another protein or to optimize detection of the first protein.
NEW Thermo Scientific NanoDrop
Catalogue No Description Pkg. Size Pay Now!
pie-34078 SuperSignal West PICO Chemiluminescent Substrate 4x1000 ml 13.980,-
pie-34080 SuperSignal West PICO Chemiluminescent Substrate 4x500 ml 8.640,-
pie-34075 SuperSignal West DURA Extended Duration Substrate 4x100 ml 12.795,-
pie-34076 SuperSignal West DURA Extended Duration Substrate 4x200 ml 20.850,-
pie-34095 SuperSignal West FEMTO Max. Sensitivity Substrate 4x100 ml 14.235,-
pie-34096 SuperSignal West FEMTO Max. Sensitivity Substrate 4x200 ml 22.500,-
Thermo Scientific CL-Xposure Film
Thermo Scientific CL-XPosure Film is an economically priced clear-blue X-ray film that exhibits the same detection sensitivity as other commercially available film brands. CL-XPosure Film works with chemiluminescent reagents for either alkaline phosphatase (AP) or horseradish peroxidase (HRP) and can also detect radioactivity, making it ideal for Western, Southern and Northern blots and Electrophoretic Mobility Shift Assays (EMSA). Make us your one-stop source for all your Western blotting or immunoblotting needs — order your substrates and film at the same time!
NEW Thermo Scientific NanoDrop
Catalogue No Description Pkg. Size Pay Now!
pie-32106 Pierce ECL Western Blotting Substrate 4x500 ml 6.615,-
pie-32209 Pierce ECL Western Blotting Substrate 4x250 ml 5.175,-
NEW Thermo Scientific Pierce Protein Gels
Thermo Scientific Pierce Protein Gels use a special formulation to produce stronger, more resilient gels, making handling during and after electrophoresis easier. The extra stability of Pierce Protein Gels combined with the Tris-HEPES-SDS Running Buffer offers both speed and excellent resolution. Compatible Gel Tanks: Thermo Scientific Owl P82 System, Novex® XCell I, II™ and Surelock® Systems, C.B.S. Scientific CBDCX-700 Dual Cool System and PAGEr® Minigel Chamber.
NEW Thermo Scientific NanoDrop
Catalogue No Description Pkg. Size Pay Now!
pie-34089 CL-XPosure Film (18 x 24 cm) 4x100 pack 6.870,-
pie-34090 CL-XPosure Film (13 x 18 cm) 4x100 pack 3.975,-
pie-34091 CL-XPosure Film (20 x 25 cm) 4x100 pack 7.815,-
pie-34092 CL-XPosure Film (13 x 18 cm) 4x25 pack 1.650,-
pie-34093 CL-XPosure Film (20 x 25 cm) 4x50 pack 4.410,-
pie-34097 CL-XPosure Film (24 x 30 cm) 4x100 pack 7.920,-
pie-34099 CL-XPosure Film (35 x 40 cm) 4x100 pack 15.825,-
pie-21065 Pierce Background Eliminator 4x1 kit 4.800,-
Thermo Scientific Restore and Restore PLUS Western Blot Stripping Buffer
Thermo Scientific Restore Western Blot Stripping Buffers are robust but gentle Western blotting stripping buffers. Restore PLUS Western Blot Stripping Buffer is a second formulation especially for high-affinity antibodies that require special treatment.
NEW Thermo Scientific NanoDrop
Catalogue No Description Pkg. Size Pay Now!
pie-21063 Restore Western Blot Stripping Buffer 5L 5.801,-
pie-46431 Restore PLUS Western Blot Stripping Buffer 5L 5.801,-
Thermo Scientific Pierce Protein Assay Kits
The Thermo Scientific Pierce 660 nm Protein Assay is a quick, ready-to-use colorimetric method for total protein quantitation. The assay is more linear than coomassie-based Bradford assays and compatible with higher concentrations of most detergents, reducing agents and other commonly used reagents.
NEW Thermo Scientific NanoDrop
Catalogue No Description Pkg. Size Pay Now!
pie-22660 Pierce 660 nm Protein Assay Reagent 2x750 ml 2.288,-
pie-22662 Pierce 660 nm Protein Assay Kit 2xkit 3.135,-
pie-22663 Ionic Detergent Compatibility Reagent (IDCR) 2 x 5x1g 1.545,-
pie-23225 Pierce BCA Protein Assay Kit 2xkit 3.398,-
pie-23227 Pierce BCA Protein Assay Kit 2xkit 2.153,-
Thermo Scientific Pierce Protein Concentrators
Thermo Scientific Pierce Protein Concentrators effectively combine simplicity, speed, capacity and recovery to yield enhanced concentration, purification and separation of proteins – even with dilute protein samples. The concentrators provide efficient concentration of biological samples containing proteins (antigens, antibodies and enzymes), nucleic acids (DNA), and microorganisms (viral particles).
NEW Thermo Scientific NanoDrop
Catalogue No Description Pkg. Size Pay Now!
pie-87748BOGO Pierce Protein Concentrators, 9K MWCO, 7 ml 20 pack 845,-
pie-87749BOGO Pierce Protein Concentrators, 9K MWCO, 20 ml 20 pack 1.515,-
pie-87750BOGO Pierce Protein Concentrators, 20K MWCO, 7 ml 20 pack 845,-
pie-87751BOGO Pierce Protein Concentrators, 20K MWCO, 20 ml 20 pack 1.515,-
pie-89920BOGO Pierce Protein Concentrators, 150K MWCO, 7 ml 20 pack 820,-
pie-89921BOGO Pierce Protein Concentrators, 150K MWCO, 20 ml 20 pack 1.475,-