Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Universal Solvent Jet NMR Tube Washer
Cleans NMR tubes in small batches. Bel-Art SP Scienceware™ Universal Solvent Jet NMR Tube Washer can also be used for cleaning gas-tight sample tubes.
Brand:  Bel-Art™ 66000-0086
Additional Details : Weight : 0.21540kg
Product Code. 15348668
  • 2795.00 DKK / Each
Estimated Shipment: 29-08-2024
to see stock.
Add to basket
Description
Description
  • Loosen and retighten the fitting on the assembled washer head to extend the PTFE tubing into the sample tube.
  • Washer head affixes to a filter flask that includes a vacuum sidearm (sold separately)
  • When a vacuum is applied to the flask, the sample tube is pressed against the rubber gasket to form an air-tight seal that starts the solvent flow.
  • Sample tube can be air-dried by lifting the PTFE tubing out of the solvent reservoir
Specifications
Specifications
Single Tube Washer
Fits tubing: 2.5 to 5.0mm O.D.
Washer Body, rubber gasket, PTFE tubing, and PTFE tubing adapter.
WG-7200-1