Learn More
DWK Life Sciences Wheaton™ Scrubber Absorber Tube
For arsenic analysis using silver diethyldithiocarbamate (SDDC) colorimetric method
Brand:  DWK Life Sciences 180034
741.00 DKK valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 0.45360kg
Product Code. 12445482
  • 780.00 DKK / Each
Estimated Shipment: 08-10-2024
to see stock.
Add to basket
Description
Description
  • Design eliminates the ball and socket joint connecting the scrubber and absorber components
  • Assembled apparatus is freestanding and easy to use
  • Manufactured from low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
Applications:
For arsenic analysis
Specifications
Specifications
Scrubber/Absorber Tube
For arsenic analysis using silver diethyldithiocarbamate method
Borosilicate Glass
1/Cs.