Learn More
Cytiva Whatman™ Grade 2668 Chr Cellulose Chromatography Paper
2668 Chr cellulose chromatography sheets is a 0.9 mm paper for separation relatively large molecules by electrophoresis.
Brand: Cytiva 10382461
6422.00 DKK valid until 2024-12-31
Use promo code "23266" to get your promotional price.
Additional Details : Weight : 10.68000kg
Description
- Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
- Manufactured and tested specifically for chromatographic techniques. This ensures the wicking capability and uniformity of capillary action that are important in chemical separations.
- Also widely used in protein and nucleic acid blotting.
Specifications
58 x 60 cm (22.8 x 23.6 in.) | |
2668 Chr cellulose chromatography sheets is a 0.9 mm paper for separation relatively large molecules by electrophoresis. | |
Grade 2668 Chr |