Learn More
R&L Enterprises™ Ward Pattern Furnace Tongs
For protection and safety when handling heated objects in the laboratory. R&L Enterprises™ Ward Pattern Furnace Tongs are fitted with a circular shield to protect the hand against radiation from the furnace.
1075.00 DKK - 1385.00 DKK
Specifications
Material | Stainless Steel |
---|---|
Type | Furnace Tongs |
For Use With (Application) | For handling a wide variety of crucibles and to protect the hand against radiation from the furnace |
Quantity | 1 Ea. |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
11869701
|
R&L Enterprises™
5705 |
740 mm |
1385.00 DKK
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
11899601
|
R&L Enterprises™
5704 |
590 mm |
1075.00 DKK
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
Description
- Manufactured from stainless steel
- For handling a wide variety of crucibles, especially in muffle furnaces
- Operated by thumb action with one hand
Specifications
Stainless Steel | |
Furnace Tongs | |
For handling a wide variety of crucibles and to protect the hand against radiation from the furnace | |
1 Ea. |