Learn More
Integra Biosciences™ Vacusafe Tubing Set with Filter
Use this filter and tubing set for the aspiration of biological liquid waste. Integra Biosciences™ Vacusafe Tubing Set with Filter is designed for use with VACUSAFE Aspiration Systems.
Brand: Integra Biosciences™ 158342
Additional Details : Weight : 0.11250kg
Description
- Silicone tubing set with hydrophobic filter
- Includes: Tubing for connection of hand operator to bottle, 180 cm, silicone (158 332); tubing for connection of instrument to bottle, 70 cm, silicone (158 331); filter for protection of vacuum source, 0.45 μm, hydrophobic (158 015)
- For use with VACUSAFE Aspiration Systems
Specifications
VACUSAFE Aspiration Systems | |
Aspiration of biological liquid waste | |
Tubing for connection of hand operator to bottle, 180 cm, silicone (Mfr. No. 158 332); tubing for connection of instrument to bottle, 70 cm, silicone (Mfr. No. 158 331); filter for protection of vacuum source, 0.45 μm, hydrophobic (Mfr. No. 158 015) |