Learn More
Heidolph instruments Unimax 2010 Circular Bews
Large model that accepts a load capacity of 10 kg for high samples throughput that features a shaking orbit of 20 mm that performs a gentle motion for sensitive samples and supports culture plates and Erlenmeyer flasks.
Brand:  Heidolph instruments 542-10020-00
17632.77 DKK valid until 2024-12-31
Use promo code "21910" to get your promotional price.
Additional Details : Weight : 14.48000kg
Product Code. 10798781
  • 19915.00 DKK / Each
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Description
Description
  • Large model that accepts a load capacity of 10 kg for high samples throughput
  • The shaking orbit of 20 mm performs a gentle motion for sensitive samples and supports culture plates and Erlenmeyer flasks
  • Set and continuously adjust the variable speed on the digital display from 20 to 400 rpm
  • Increase your sample throughput with the optional multi-tier attachment for a variety of vessel sizes
  • A digital process timer allows for unattended operation and can be set from 1 to 120 minutes
Specifications
Specifications
10 mm
30 to 500 rpm
230 V
Unimax 2010 Circular Bew.
5 kg
230 V
Shaker