Learn More
Retsch Ultra Centrifugal Mill ZM 300
Rotor mills are used for high-speed size reduction of soft to medium-hard as well as temperature-sensitive or fibrous materials.
Brand: Retsch 20.824.0001
70903.25 DKK valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 36.00000kg
Description
- The powerful Ultra Centrifugal Mill ZM 300 provides maximum grinding performance combined with ease of use.The variable speed from 6000 to 23,000 rpm allows for gentle, neutral-to-analysis sample preparation in a very short time.
- Thanks to an integrated temperature monitoring system, reproducibility is guaranteed even for long grinding processes or pulverization of large sample volumes.
- The wide selection of rotors, ring sieves and cassettes makes the ZM 300 a true allrounder which meets the requirements of a great variety of size reduction tasks.
Specifications
Ultra-Centrifugal Mill | |
900 mL (standard cassette); 1200 mL (volume cassette); 5000 mL / 3000 mL / 500 mL / 250 mL (cyclone) | |
426 x 452 x 431 mm | |
Agriculture, biology, chemistry/plastics, construction materials, engineering/electronics, environment/recycling, food, geology/metallurgy, medicine/pharmaceuticals | |
1-phase | |
100/120 V or 200/240 V | |
452 mm |
Not Autoclavable | |
CE | |
No | |
IP20 | |
6000 to 23,000 min-1, free selectable | |
38 kg |