Learn More
Cole-Parmer™ LaMotte™ Secchi Turbidity Disk
Use this disk to determine turbidity or degree of visibility in natural water supplies. Cole-Parmer™ LaMotte™ Secchi Turbidity Disk features braided, stretch resistant lines marked at half and full meter intervals up to 20 meters.
Brand:  Cole-Parmer™ 05492-00
Additional Details : Weight : 0.74700kg
Product Code. 11762373
  • 845.00 DKK / Each
Estimated Shipment: 18-07-2024
to see stock.
Add to basket
Description
Description
Simple-to-use secchi disk is a weighted, 20-cm dia. black and white plate that sinks parallel to the water's surface. For best results use from the leeward side of a boat or dock between mid-morning and mid-afternoon.
Specifications
Specifications
Turbidity Disk
1 Ea.