Learn More
TST™ Bowie Dick™ Test Pack
Test the steam penetration and air removal capability of a vacuum sterilizer with this test pack. The TST™ Bowie Dick Test Pack is a unique combination of advance Browne intelligent ink technology and the original Bowie Dick concept.
1950.00 DKK - 4045.00 DKK
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | ISO 11140-4(3) Class 2 |
For Use With (Application) | For large porous load sterilizers |
Product Type | TST Bowie Dick Test Pack |
Sterilization Indicator | Yes |
Product Code | Brand | Sterilization Method | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterilization Method | Price | Quantity & Availability | |||||
12390489
|
Steris Corporation
2310AB |
Steam, Vacuum |
1950.00 DKK
Pack of 20 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
12380489
|
Steris Corporation
2352AB |
Steam |
4045.00 DKK
Pack of 20 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
Description
- Single-use test pack
- A successful test confirms that steam penetration into a test pack is rapid and even and that air and other non-condensable gases have been effectively removed
- The chemical indicator sheet at the center of the pack shows a defined color change from yellow to dark blue/purple when exposed to a specific combination of time, temperature and steam
- Conforms to ISO 11140-4 Class 2
Specifications
Autoclavable | |
For large porous load sterilizers | |
Yes |
ISO 11140-4(3) Class 2 | |
TST Bowie Dick Test Pack |