Learn More
Sampling Systems™ Stainless Steel Tweezers
These stainless steel tweezers are essential equipment for laboratories.
17.50 DKK - 40.70 DKK
Specifications
Material | Stainless Steel (304) |
---|---|
Product Type | Tweezers |
For Use With (Application) | General laboratory use |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
15809651
|
Sampling Systems™
A394-100 |
100 mm |
17.50 DKK
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
15819651
|
Sampling Systems™
A394-200 |
200 mm |
40.70 DKK
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
- Polished finish
- Two sizes available
- Made from hygienic 304 grade stainless steel
Specifications
Stainless Steel (304) | |
Tweezers | |
General laboratory use |