Learn More
Bochem™ Stainless Steel Transport Basket
Brand:  Bochem™ 17306
Additional Details : Weight : 0.50000kg
Product Code. 15395803
  • 3205.00 DKK / Each
Estimated Shipment: 01-07-2024
to see stock.
Specifications
18/10 E-Poli
600 x 400 x 150 mm
400 mm
Yes
600 mm
150 mm