Learn More
Nabertherm™ Split Type Horizontal Tube Furnace RSH 1100 Series with P580 Controller
The split-type design makes it easy to change the working tube. It allows for a comfortable exchange of various working tubes.
52015.41 DKK - 97771.09 DKK
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Length (Metric) | Price | Quantity & Availability | ||||
17328159
|
Nabertherm™
RS252W2AN3 |
50 mm | 500 mm |
52015.41 DKK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17368159
|
Nabertherm™
RS322W2AN3 |
120 mm | 500 mm |
63715.15 DKK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17308169
|
Nabertherm™
RS382W2AN3 |
80 mm | 500 mm |
61852.14 DKK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17348169
|
Nabertherm™
RS422W2AN3 |
120 mm | 750 mm |
79662.57 DKK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17388169
|
Nabertherm™
RS472W2AN3 |
170 mm | 750 mm |
88679.57 DKK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17328179
|
Nabertherm™
RS482W2AN3 |
80 mm | 750 mm |
76309.14 DKK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17368179
|
Nabertherm™
RS522W2AN3 |
120 mm | 1000 mm |
85400.66 DKK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
17308189
|
Nabertherm™
RS572W2AN3 |
170 mm | 1000 mm |
97771.09 DKK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||||
Description
- Maximum Temperature: 1100°C
- Dual shell housing made of sheets of textured stainless steel
- Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
- Type N thermocouple
- Split-type design for simple insertion of the working tube
- Working tube made of material C 530 including fiber plugs for operation under air in scope of delivery
- Heating elements on support tubes provide for free radiation
- Switchgear and control unit integrated in furnace housing
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Specifications
P580 | |
Up to 1100°C |