Learn More
Azpack™ Spare Kit for AP1/AP2 Aquarium Pumps
Replace worn out and damaged diaphragms, valves, and gaskets for AP1 and AP2 Aquarium Pumps. Azpack™ Spare Kit for AP1/AP2 Aquarium Pumps contains all the necessary parts for tank maintenance including a handy tool to get the job done.
Brand: Azpack™ 12403180
Additional Details : Weight : 0.01000kg
Description
Kit includes:- 1 diaphragm
- 2 valve flappers
- 2 valve retainers
- 1 gasket
- 1 pivot plug
- 1 handy screwdriver/spanner
- 1 air filter pad
- Instructions for use and installation
Specifications
AP1 and AP2 Aquarium Pumps | |
1 diaphragm, 2 valve flappers, 2 valve retainers,1 gasket, 1 pivot plug,1 handy screwdriver/spanner, 1 air filter pad, and instructions. | |
1 Each |