Learn More
Brady™ Safety Sign COVID-19 Reception2
An efficient solution with clear safety instructions to easily communicate COVID-19 countermeasures on work floors.
144.00 DKK - 148.00 DKK
Specifications
Material | Laminated Polyester |
---|---|
Mounting Type | Self-adhesive |
Dimensions (L x W) | 262 x 371 mm (W x H) |
Product Code | Brand | Languages | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Languages | Price | Quantity & Availability | |||||
16506434
|
Brady™
306839 |
Finnish |
146.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
16516434
|
Brady™
306840 |
Swedish |
146.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
16526404
|
Brady™
306811 |
Dutch |
144.00 DKK
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
16526444
|
Brady™
306851 |
Norwegian |
146.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
16536404
|
Brady™
306812 |
French |
148.00 DKK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16546404
|
Brady™
306813 |
German |
148.00 DKK
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||||
16556404
|
Brady™
306814 |
English |
148.00 DKK
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
16566404
|
Brady™
306815 |
Spanish |
146.00 DKK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16576404
|
Brady™
306816 |
Italian |
146.00 DKK
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16596424
|
Brady™
306838 |
Danish |
146.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
Description
- Provides clear, visual communication where it matters
- Highlights the use of appropriate personal protection
- Easily communicates social distancing and hygiene
Specifications
Laminated Polyester | |
Self-adhesive | |
262 x 371 mm (W x H) |