Learn More
3M™ Safety Glasses Microfiber Pouch
170.00 DKK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Use promo code "22912" to get your promotional price.
Click to view available options
Size: 11 x 19 cm
Description
- Microfiber pouch to protect safety glasses
- Helps cleaning your 3M™ Safety Glasses
Specifications
Dimensions (L x W) | 11 x 19 cm |
Product Type | Eyewear bag |
For Use With (Equipment) | Safety Glasses |
Material | Microfiber |