Learn More
Epredia™ HistoGel™ Specimen Processing Gel
Encapsulate and retain even small, friable and viscous specimens during histological processing with HistoGel™ Specimen Processing Gel.
3015.00 DKK - 9825.00 DKK
Product Code | Brand | Product Type | Content And Storage | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Content And Storage | Price | Quantity & Availability | ||||
12006679
|
Epredia™
HG-4000-012 |
Specimen Processing Gel |
3015.00 DKK
Pack of 12 |
In Stock
Log in to see stock. |
|||||
12608606
|
Epredia™
HG-4000-144 |
Specimen Processing Gel |
8820.00 DKK
Pack of 144 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
12056689
|
Epredia™
HG-4000-144 |
Specimen Processing Gel |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Penetrates specimen without allowing specimen to escape from gel
- Outperforms other agarose gel mixtures or thromboplastin at a fraction of the cost
- Eliminates unwanted discoloration around specimens on slides
- Remains inside theparaffin block during cutting and virtually unnoticeable during sectioning
Specifications
Specimen Processing Gel |