Learn More
RETSCH Test Sieves - 14 Mesh or Greater
Stainless steel with wire gauze
915.00 DKK - 6405.00 DKK
Specifications
Mesh Material | Stainless Steel |
---|---|
For Use With (Application) | Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc. |
Depth (English) | 2 in. |
Depth (Metric) | 5.08cm |
Type | Test Sieve |
Product Code | Brand | Pore Size | Tyler Standard | U.S. Standard | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Tyler Standard | U.S. Standard | Price | Quantity & Availability | |||
10014791
|
RETSCH
60.150.000020 |
20μm | 635 Mesh | No. 635 |
6405.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10790463
|
RETSCH
60.150.000025 |
25μm | 500 Mesh | No. 500 |
3370.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10390902
|
RETSCH
60.150.000032 |
32μm | 450 Mesh | No. 450 |
1950.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10468341
|
RETSCH
60.150.000038 |
38μm | 400 Mesh | No. 400 |
1950.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10534492
|
RETSCH
60.150.000045 |
45μm | 325 Mesh | No. 325 |
1345.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10248971
|
RETSCH
60.150.000053 |
53μm | 270 Mesh | No. 270 |
1180.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10534682
|
RETSCH
60.150.000075 |
75μm | 200 Mesh | No. 200 |
915.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10045071
|
RETSCH
60.150.000106 |
106μm | 140 Mesh | No. 140 |
915.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10085031
|
RETSCH
60.150.000063 |
63μm | 230 Mesh | No. 230 |
915.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10791423
|
RETSCH
60.150.000090 |
90μm | 170 Mesh | No. 170 |
915.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10701433
|
RETSCH
60.150.000125 |
125μm | 120 Mesh | No. 120 |
915.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10752003
|
RETSCH
60.150.000212 |
212μm | 70 Mesh | No. 70 |
915.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10585062
|
RETSCH
60.150.000150 |
150μm | 100 Mesh | No. 100 |
915.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10004921
|
RETSCH
60.150.000180 |
180μm | 80 Mesh | No. 80 |
915.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10555642
|
RETSCH
60.150.000250 |
250μm | 60 Mesh | No. 60 |
915.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10610092
|
RETSCH
60.150.000300 |
300μm | 50 Mesh | No. 50 |
915.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10278541
|
RETSCH
60.150.000425 |
425μm | 40 Mesh | No. 40 |
915.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10024931
|
RETSCH
60.150.000355 |
355μm | 45 Mesh | No. 45 |
915.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10238591
|
RETSCH
60.150.000500 |
500μm | 35 Mesh | No. 35 |
915.00 DKK
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
10523342
|
RETSCH
60.150.000710 |
710μm | 25 Mesh | No. 25 |
935.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10620092
|
RETSCH
60.150.000600 |
600μm | 30 Mesh | No. 30 |
935.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10208691
|
RETSCH
60.150.000850 |
850μm | 20 Mesh | No. 20 |
935.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10330622
|
RETSCH
60.150.001000 |
1mm | 18 Mesh | No. 18 |
965.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10340622
|
RETSCH
60.150.001180 |
1.18mm | 16 Mesh | No. 16 |
965.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10583722
|
RETSCH
60.150.001400 |
1.4mm | 14 Mesh | No. 14 |
965.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
10136651
|
RETSCH
60.150.025000 |
25mm | 710 Mesh | 710 |
1325.00 DKK
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||
Specifications
Stainless Steel | |
2 in. | |
Test Sieve | |
8 in. |
Ideal for cement clinker, chemicals, coffee, construction materials, fertilizers, fillers, flours, grains, Metals powders, Minerals, nuts, Plastics, sand, seeds, soils, washing powder etc. | |
5.08cm | |
ASTM E11 | |
Stainless Steel |