Learn More
Pyrex™ Short Stem Glass Funnel
Manufactured from chemical and heat resistant Pyrex™ Borosilicate Glass. Pyrex™ Short Stem Glass Funnel with 60° bowl angle provides precise fitting of filter paper.
102.42 DKK - 1645.00 DKK
Specifications
Material | Pyrex Glass, Borosilicate Glass |
---|---|
Certifications/Compliance | NF B 35 032 |
Product Type | Short stem funnel |
Product Code | Brand | Diameter (Metric)Top | Height (Metric) | Diameter (Metric) Disc | Length (Metric) Stem | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Height (Metric) | Diameter (Metric) Disc | Length (Metric) Stem | Price | Quantity & Availability | ||
12529945
|
Pyrex
2140/03M |
70mm | 125 mm | 70 to 90mm (Filter paper size) | 75mm |
1430.00 DKK
Pack of 10 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||
12595067
|
Pyrex
2140/06M |
80mm | 144 mm | 90 to 150mm (Filter paper size) | 82mm |
1645.00 DKK
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||
12099907
|
Pyrex
2140/11M |
125mm | 220 mm | 185 to 240mm (Filter paper size) | 122mm |
198.00 DKK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||
11307944
|
Pyrex
2140/14M |
200mm | 345 mm | 320 to 385mm (Filter paper size) | 193mm |
665.00 DKK
Each |
Estimated Shipment: 29-08-2024
Log in to see stock. |
||
11397934
|
Pyrex
2140/12M |
150mm | 270 mm | 240 to 270mm (Filter paper size) | 154mm |
387.00 DKK
Each |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||
12585067
|
Pyrex
2140/01M |
35mm | 60 mm | 45 to 55mm (Filter paper size) | 35mm |
1230.00 DKK
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||
11347934
|
Pyrex
2140/02M |
45mm | 78 mm | 55 to 70mm (Filter paper size) | 45mm |
1155.00 DKK
Pack of 10 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||
11992568
|
Pyrex
2140/10M |
105mm | 186 mm | 150 to 185mm (Filter paper size) | 106mm |
1500.00 DKK
Pack of 10 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||
11357934
|
Pyrex
2140/03M |
70mm | 125 mm | 70 to 90mm (Filter paper size) | 75mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12923611
|
Pyrex
2140/02M |
45mm | 78 mm | 55 to 70mm (Filter paper size) | 45mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11367934
|
Pyrex
2140/06M |
80mm | 144 mm | 90 to 150mm (Filter paper size) | 82mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11337934
|
Pyrex
2140/01M |
35mm | 60 mm | 45 to 55mm (Filter paper size) | 35mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Manufactured from chemical and heat resistant Pyrex™ Borosilicate Glass. Pyrex™ Short Stem Glass Funnel with 60° bowl angle provides precise fitting of filter paper.
- With 60° bowl angle for precise fitting of filter paper
- Manufactured from chemical and heat resistant Pyrex™ borosilicate glass
Specifications
Pyrex Glass, Borosilicate Glass | |
NF B 35 032 | |
Short stem funnel |