Learn More
Pyrex™ Borosilicate Glass Narrow Neck Flat Bottom Boiling Flask
Manufactured from Pyrex™ glass for high resistance to thermal shock. Pyrex™ Borosilicate Glass Narrow Neck Flat Bottom Boiling Flask complies with ISO 1773 standards. Excellent chemical durability for contamination-free performance. Includes white enamel marking spot while flat base prevents spillage.
89.05 DKK - 10575.00 DKK
Specifications
Certifications/Compliance | ISO 1773 |
---|---|
Material | Borosilicate Glass, Borosilicate Glass |
Neck Style | Narrow |
Bottom Shape | Flat |
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Diameter (Metric) Inner Neck | Height (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Diameter (Metric) Outer | Diameter (Metric) Inner Neck | Height (Metric) | Price | Quantity & Availability | ||
10650191
|
Pyrex
1070/38D |
10 L | 280 mm | 57 mm | 360 mm |
1905.00 DKK
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||
10009569
|
Pyrex
1070/04D |
100 mL | 64 mm | 18 mm | 110 mm |
89.05 DKK
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||
10662111
|
Pyrex
1070/42D |
20 L | 341 mm | 60 mm | 558 mm |
3565.00 DKK
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||
10801601
|
Pyrex
1070/28D |
2000 mL | 166 mm | 45 mm | 250 mm |
3240.00 DKK
Pack of 10 |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||
10650001
|
Pyrex
1070/36D |
6 L | 237 mm | 57 mm | 340 mm |
1110.00 DKK
Each |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||
12079002
|
Pyrex
1070/12D |
250 mL | 85 mm | 30 mm | 140 mm |
900.00 DKK
Pack of 10 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||
12089002
|
Pyrex
1070/18D |
500 mL | 105 mm | 30 mm | 170 mm |
10575.00 DKK
Pack of 10 |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||
11982609
|
Pyrex
1070/24D |
1000 mL | 131 mm | 36 mm | 200 mm |
1815.00 DKK
Pack of 10 |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||
10640001
|
Pyrex
1070/32D |
4 L | 207 mm | 45 mm | 300 mm |
845.00 DKK
Each |
Estimated Shipment: 05-07-2024
Log in to see stock. |
||
12549085
|
Pyrex
1070/02M |
50 mL | 50 mm | 18 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10211560
|
Pyrex
1070/40D |
12 L | 300 mm | 57 mm | 465 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
- Complies with ISO 1773
- Flat base for standing on benches and hotplates etc
- Manufactured from Pyrex™ glass for high resistance to thermal shock
- Excellent chemical durability for contamination free performance
- With white enamel marking spot
Specifications
ISO 1773 | |
Borosilicate Glass, Borosilicate Glass | |
Narrow | |
Flat |