missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™120mm Diameter Glass Büchner Filter Funnel
Manufactured from chemically resistant Pyrex™ glass. Pyrex™ 120mm Diameter Glass Büchner Filter Funnel with integral sintered glass disc is highly resistant to most corrosive reagents and eliminates the need for filter papers.
2375.00 DKK - 3005.00 DKK
Specifications
Capacity (Metric) | 1000 mL |
---|---|
Diameter (Metric) Stem | 19mm |
Material | Pyrex Glass, Borosilicate Glass |
Height (Metric) | 290 mm |
For Use With (Application) | For easy filtering solutions |
Product Code | Brand | Pore Size | Porosity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Pore Size | Porosity | Price | Quantity & Availability | ||||
12683222
|
Pyrex
3720/02M |
100 to 160μm | 1 |
2375.00 DKK
Pack of 2 |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||||
12693222
|
Pyrex
3720/04M |
40 to 100μm | 2 |
2375.00 DKK
Pack of 2 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
||||
11901558
|
Pyrex
3720/06M |
16 to 40μm | 3 |
2380.00 DKK
Pack of 2 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
||||
12613232
|
Pyrex
3720/08M |
10 to 16μm | 4 |
3005.00 DKK
Pack of 2 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
||||
Description
- Comprehensive range of Büchner filter funnels with integral sintered glass discs
- Disc and funnel body manufactured from chemically resistant Pyrex™ glass
- For convenient filtration of solutions
- No filter papers required
- Highly resistant to most corrosive reagents including ammonia and concentrated sulphuric acid - Choice of disc diameters and porosities
Specifications
1000 mL | |
Pyrex Glass, Borosilicate Glass | |
For easy filtering solutions | |
Integrated filter plate sintered Glass disc | |
Buchner filter funnel |
19mm | |
290 mm | |
120mm | |
120mm |