Learn More
Haldenwanger™ Porcelain Evaporating Dishes
Designed with glazed laboratory porcelain to provide excellent thermal and chemical resistance. Haldenwanger™ Porcelain Evaporating Dishes provide a solid basin for evaporating excessive solvents to produce a concentrated solution or solid precipitate.
64.77 DKK - 212.00 DKK
Specifications
Material | Porcelain |
---|---|
Shape | Round |
Type | Evaporating Dish |
For Use With (Application) | Laboratories |
Interior Finish | Glazed |
Product Code | Brand | Bottom Shape | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Bottom Shape | Diameter (Metric) | Height (Metric) | Price | Quantity & Availability | |||
10494531
|
Haldenwanger™
888/4 |
Flat | 100 mm | 20 mm |
147.00 DKK
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||
10260011
|
Haldenwanger™
888/2 |
Flat | 80 mm | 16 mm |
107.00 DKK
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
10483381
|
Haldenwanger™
109/5 |
Round | 125 mm | 50 mm |
212.00 DKK
Each |
In Stock
Log in to see stock. |
|||
10269530
|
Haldenwanger™
109/000 |
Round | 63 mm | 25 mm |
64.77 DKK
Each |
In Stock
Log in to see stock. |
|||
10128160
|
Haldenwanger™
109/00 |
Round | 70 mm | 28 mm |
81.30 DKK
Each |
In Stock
Log in to see stock. |
|||
10443001
|
Haldenwanger™
109/0 |
Round | 80 mm | 32 mm |
92.06 DKK
Each |
In Stock
Log in to see stock. |
|||
Description
- Available in shallow and semi-shallow form
- Sizes up to 175mL glazed; 1,000mL and larger are glazed except outside base
Specifications
Porcelain | |
Evaporating Dish | |
Glazed |
Round | |
Laboratories |