Learn More
Alpha Packaging™ Polyethylene Wallet Bag
Brand: Alpha Packaging™ 12330559
Additional Details : Weight : 0.00900kg
Specifications
Wallet Bag | |
Green | |
Polyethylene | |
25 | |
485 x 340mm | |
340 mm |
Zip Seal | |
Useful for filling and carrying documents, presentations and storage | |
125 μm | |
25 Pack | |
485 mm |