Learn More
3M™ PELTOR™ Rechargeable NimH Battery Pack ACK03
3M™ PELTOR™ Rechargeable NimH Battery Pack ACK03 designed for use with 3M™ PELTOR™ Alert, LiteCom, WS™, Tactical XP, FM Radio and Radio XP headsets.
Brand:  3M™ 7100064601
174.00 DKK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 4.98000kg
Product Code. 15790078
  • 281.00 DKK / Each
Estimated Shipment: 18-07-2024
to see stock.
Description
Description
  • Rechargeable NiMH battery pack, 1700 mAh
  • For use with 3M™ PELTOR™ Alert, LiteCom, WS™, Tactical XP, FM Radio and Radio XP Headsets
  • PELTOR™ FR03 charger supplied separately
  • 1700 mha
  • Charging time 5-6 hours
  • NiMh Battery
Specifications
Specifications
Battery Pack
3M™ PELTOR™ Alert, LiteCom, WS™, Tactical XP, FM Radio and Radio XP Headsets.