Learn More
3M™ PELTOR™ Rechargeable Li-ion Battery Pack
3M™ PELTOR™ Rechargeable Li-ion Battery Pack designed for use with 3M™ PELTOR™ LiteCom Plus Headsets.
Brand:  3M™ 7100075380
758.00 DKK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 6.33101kg Transport : UN number : 3480 Chem class : 9
Product Code. 15740088
  • 1060.00 DKK / Each
Estimated Shipment: 17-07-2024
to see stock.
Description
Description
  • Rechargeable Lithium-Ion battery pack
  • Battery Capacity 3.7 V, 1350mAh, 5 Wh
  • For use with 3M™ PELTOR™ LiteCom Plus headsets
  • Charging time 4 hours
Specifications
Specifications
Battery Pack