Learn More
3M™ PELTOR™ CH-3 Listen Only Headset
PELTOR™ CH-3 Listen Only Headsets utilise the FLX2 cable system for a simple and reliable connection. They feature a speaker system that’s optimised for speech frequency in high noise environments and a modern cup design which provides high comfort and attenuation.
16395.00 DKK
Especificaciones
Color | Blue |
---|---|
Product Type | Communication Headset |
Noise Level | 25 dB |
Código de producto | Marca | For Use With (Equipment) | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | For Use With (Equipment) | Precio | Cantidad y disponibilidad | |||||
17879066
|
3M™
7100203297 |
Hard Hat |
16395.00 DKK
Pack of 10 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
|||||
17889066
|
3M™
7100203639 |
16395.00 DKK
Pack of 10 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||||||
Descripción
- Protect your hearing without losing the ability to hear conversation with PELTOR™ CH-3 Listen Only Headsets.
- These hear-only headsets feature a CH-3 speaker system that’s optimized for the speech frequency in high noise environments and meets the efficiency demand from today's digital devices.
- The FLX2 cable system provides a simple and reliable connection and the modern cup design offers high comfort and attenuation.
- All critical parts are rigorously tested and easy to change without any special tooling which contributes to low maintenance costs.
- Comes with a headband, neckband or helmet attachment.
- Available in bright yellow for the standard model or bright blue to indicate Explosive atmosphere (Ex) approval.
Especificaciones
Blue | |
Communication Headset | |
25 dB |