Learn More
Thermo Scientific™ Cell Locker™ Chamber with Copper Tray and Transport Cover
Protect your most sensitive cells, even during transport.
Brand:  Thermo Scientific™ 50154739X6
Additional Details : Weight : 11.00000kg
Product Code. 15804295
  • 54847.20 DKK / Each
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
Protect your most sensitive cells, even during transport.
Each individual Cell Locker™ Chamber is effectively a quarantine chamber. Can be manually disinfected or can be autoclaved.
  • Dual 0.2 μm membrane filters permit air circulation but exclude microbial contaminants
  • Replacable 0.2 μm membrane filter is hydrophobic, oleophobic, resistant to organic solvents and tested for biosafety and low cytotoxicity
  • Dishes and flasks can be removed individually on a work tray or inside the entire chamber with optional transport cover, thus protecting samples from outside air
  • Non-skid tray slides out to allow access to culture vessels, or the entire tray can be removed
Specifications
Specifications
Cell Protection and Transport
Chamber, Copper Tray, and Transport Cover
6 Pk.