missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Normal Rabbit Serum

Applications: Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Control, Immunocytochemistry, Block/Neutralize

Brand:  Novus Biologicals™ NBP1-71681

Additional Details : Weight : 0.00970kg

Product Code. 15746087

  • 3022.50 DKK / 5mL
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Host: Rabbit
  • Contains 0.1% Sodium Azide as presevative
Applications: 

Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry-Paraffin

Recommended Storage

Store at 4°C for short term and -20°C for long term

Specifications

Specifications

Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Control, Block/Neutralize, Fluorescence Imaging
5mL
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Control, Block/Neutralize, Fluorescence Imaging
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers