Learn More
Fisherbrand™ Bunsen Safety Burner for LP Gas
Supplied with a flame failure device that shuts off the gas supply to the burner within approximately 20 seconds of the flame being extinguished.
Brand:  Fisherbrand™
Additional Details : Weight : 0.39600kg
Product Code. 12862144
  • 585.00 DKK / Each
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
Description
Description
  • Safety features all contained within the burner
  • For use with natural gas/methane
  • 12.7mm diameter opening
Specifications
Specifications
Natural Gas, Methane
1 Ea.