Learn More
Thermo Scientific™ White Light Conversion Screen for myECL™ Imager
For use with Thermo Scientific myECL Imager
Marke: Thermo Scientific™ 62243
Additional Details : Weight : 0.50000kg
Beschreibung
Thermo Scientific™ myECL Imager Accessories and Replacement Parts are for use with Thermo Scientific myECL Imager.
Replacement parts for the myECL Imager and accessories to expand upon its capabilities.
Spezifikation
24.5 cm x 19.5 cm | |
White Light Conversion Screen | |
White Light Conversion Screen |
myECL Imager | |
myECL |