missing translation for 'onlineSavingsMsg'
Learn More

Myc Tag Monoclonal Antibody, HRP, Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen R95125

Additional Details : Weight : 0.00525kg

Product Code. 10403212

  • 2580.00 DKK / Each
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NOTE: R951-25 was previously sold under product #46-0709. When re-ordering, please use #R951-25. This antibody recognizes amino acid sequence: Glu-Gln-Lys-Leu-Ile-Ser-Glu-Glu-Asp-Leu-. This product contains enough material for 25 Western blots. Western blot against 500 ng of recombinant protein gave a strong signal using chemiluminescent or alkaline phosphatase detection reagents. For Western blot, dilute in PBS containing 0.05% Tween-20 and 5% nonfat, dry milk (PBSTM).

Myc is a proto-oncogene and encodes a nuclear phosphoprotein that plays a role in cell cycle progression, apoptosis and cellular transformation. The encoded protein forms a heterodimer with the related transcription factor MAX. This complex binds to the E box DNA consensus sequence and regulates the transcription of specific target genes. Amplification of this gene is frequently observed in numerous human cancers. Translocations involving this gene are associated with Burkitt lymphoma and multiple myeloma in human patients. There is evidence to show that translation initiates both from an upstream, in-frame non-AUG (CUG) and a downstream AUG start site, resulting in the production of two isoforms with distinct N-termini.
Specifications

Specifications

Myc Tag
Monoclonal
PBS with no preservative
P01106
MYC
c-Myc synthetic peptide: Glu-Gln-Lys-Leu-Ile-Ser-Glu-Glu-Asp-Leu-
50 μL
Primary
Tag
Antibody
IgG1
ELISA, Western Blot
HRP
MYC
46-0603; AU016757; avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; class E basic helix-loop-helix protein 39; cmyc; c-Myc; c-myc epitope tag; c-myc proto-oncogene; MRTL; MYC; Myc protein; Myc proto-oncogene protein; Myc Tag; Myc2; MYCC; myc-related translation/localization regulatory factor; myelocytomatosis oncogene; Niard; Nird; Proto-oncogene c-Myc; R95025; R950-25; transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog
Mouse
purified
RUO
4609
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Tag
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers