Learn More
Thermo Scientific™ Megafuge ST1R Plus, 208-240V TX--400 Cell Culture Package
Waste no time finding the right part numbers for your application! The application driven centrifuge package ensures convenient, fast ordering of all components necessary to run a specific protocol.
Brand:  Thermo Scientific™ 75016025
92057.06 DKK valid until 2024-04-26
Use promo code "21699" to get your promotional price.
Additional Details : Weight : 132.00000kg
Product Code. 17182449
  • POA / 1 set
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
Waste no time finding the right part numbers for your application! The application driven centrifuge package ensures convenient, fast ordering of all components necessary to run a specific protocol. Run 16 x 50 mL conicals under biocontainment certified lids. Contains centrifuge, TX-400 rotor, buckets, biocontainment lids, 50 mL conical adapters (maximum capacity 16 x 50mL), 15 mL conical adapter (maximum capacity 36 x 15 mL).
  • Easy to Use:
    • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
    • Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications
    • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.
    Thermo Scientific™ Auto-Lock™ III Rotor System:
    • Secure locking system allows easy push-button installation and exchange of rotors.
    • System saves time when switching between applications.
    • Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.
    Thermo Scientific™ ClickSeal™ Bucket Sealing System:
    • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips.
    • System has glove-friendly, one-handed open/close capability.
    • System is certified by CAMR™ in Porton Down, UK.
    Thermo Scientific™ SMARTSpin™ Technology:
    • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option:
    • Technology optimizes safety and improves separations.
    Space-Saving Design:
    • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.
    Energy Efficient:
    • System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
Specifications
Specifications
Direct, Brushless induction low profile motor with choices of 9 acceleration, 10 deceleration rates
Part Numbers In Pk. 75009770 (centrifuge),75003181(rotor),75003655(set of buckets),75003656(lids),75003682(set of adapters),75003683(set of adapters)
General Purpose Centrifuge Series
Special Offers
Services