Learn More
Kinematica Polytron™ ST-P10/600 Homogenizer Stand
Plate stand for safe and easy height adjustment of the drive unit
Brand:  Kinematica 11040050
Additional Details : Weight : 0.80000kg
Product Code. 10103943
  • 4390.00 DKK / Each
Estimated Shipment: 17-07-2024
to see stock.
Add to basket
Description
Description
  • Plate stand for Polytron PT 10-35 GT
  • Easy to use design-takes up less space  
  • Easy to clean anodized aluminum
Specifications
Specifications
Homogenizer Stand
ST-P10/600