Learn More
DWK Life Sciences Kimble™ Closures for 20mL Glass and Plastic Scintillation Vials
Durable screw thread closures
3875.00 DKK
Specifications
Color White
View More Specs
Products 1
Product Code Brand ClosureMaterial Material (Liner) Price Quantity & Availability  
Product Code Brand ClosureMaterial Material (Liner) Price Quantity & Availability  
11299819
missing translation for 'viewDocuments' Promotion Details
DWK Life Sciences
74525-22400
Polyethylene
3875.00 DKK
Pack of 1000
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
Description
Description
For 20mL Fisherbrand™ and Kimble scintillation vials, except 03-340-4B and 03-340-129. GPI 22-400 thread.