Learn More
Invitrogen™ pRSET-BFP Bacterial Expression Vector
Allow fusion of a protein of interest to the widely used and well-characterized fluorescent proteins from the jellyfish Aequorea victoria
Marca: Invitrogen™ V35420
Dettagli aggiuntivi : Weight : 0.00525kg
Descrizione
- These vectors contain the EGFP, for Emerald Green Fluorescent Protein (EmGFP), which has been further optimized for mammalian expression
- Vector also codes for Cyan (CFP) and Blue (BFP) Fluorescent Proteins for simple, noninvasive detection of recombinant protein
Bacterial Expression, Enzyme and Protein Activity Assays, Fluorescent Protein (e.g. GFP) Assays, Protein Expression, Proteins, Expression, Isolation and Analysis, Reporter Assays
Specifica
Inducible | |
T7 | |
BFP | |
Store in freezer (-5 to -30°C). | |
EK (Enterokinase) Recognition Site | |
Restriction Enzyme/MCS | |
Vivid Colors™ |
IPTG | |
Bacterial Expression Vector | |
None | |
Ampicillin (AmpR) | |
His Tag (6x), Xpress Epitope Tag | |
pRSET |
For Research Use Only. Not for use in diagnostic procedures.