Learn More
METTLER TOLEDO™ InLab™ Expert pH Electrodes
A combination pH electrode with PEEK plastic shaft for chemical and mechanical durability, and a fixed cable. Designed for use in a variety of applications.
1525.00 DKK - 4140.00 DKK
Specifications
Material | PEEK |
---|---|
pH Range | 0 to 14 |
Temperature (Metric) Operating | 0°C to 100°C |
Filling Solution Type | Xerolyt Polymer |
Internal Reference | ARGENTHAL |
Product Code | Brand | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | ||
10454353
|
METTLER TOLEDO™
51343104 |
InLab™ Expert NTC30 | Combination Electrode, Integrated Temperature Probe (NTC30) | Multi-Pin | Multi-Pin to BNC (30281896) |
2135.00 DKK
Each |
In Stock
Log in to see stock. |
||
10009552
|
METTLER TOLEDO™
51343100 |
InLab™ Expert | Combination pH Electrode | S7 | S7 to BNC (30281915) |
1525.00 DKK
Each |
In Stock
Log in to see stock. |
||
10610904
|
METTLER TOLEDO™
51343103 |
InLab™ Expert DIN | Combination pH Electrode, Integrated Pt1000 Temperature Probe | DIN 19262, 4 mm Banana Plug | pH Meter |
2210.00 DKK
Each |
In Stock
Log in to see stock. |
||
10040073
|
METTLER TOLEDO™
51343102 |
InLab™ Expert Pro-ISM (2 m) | Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology | BNC/RCA | Fixed Cable to sensor |
2895.00 DKK
Each |
In Stock
Log in to see stock. |
||
10454543
|
METTLER TOLEDO™
51343105 |
InLab™ Expert Pt1000 | Combination pH Electrode, Integrated Pt1000 Temperature Probe | Multi-Pin | Multi-Pin to BNC (30281896 |
2135.00 DKK
Each |
In Stock
Log in to see stock. |
||
10660324
|
METTLER TOLEDO™
51343101 |
InLab™ Expert Pro | Combination Electrode, Integrated Temperature Probe | BNC/RCA | Fixed Cable to sensor |
2445.00 DKK
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||
10447144
|
METTLER TOLEDO™
51340288 |
InLab™ Expert Go | Combination Electrode, Integrated Temperature Probe | BNC/RCA IP67 | Fixed Cable to sensor |
2765.00 DKK
Each |
In Stock
Log in to see stock. |
||
10431435
|
METTLER TOLEDO™
51344102 |
InLab™ Expert Go-ISM | Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology | BNC/RCA IP67 | Fixed Cable to sensor |
3540.00 DKK
Each |
In Stock
Log in to see stock. |
||
10594925
|
METTLER TOLEDO™
51344104 |
InLab™ Expert Go-ISM (10 m) | Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology | BNC/RCA IP67 | Fixed Cable to sensor |
4140.00 DKK
Each |
In Stock
Log in to see stock. |
||
11394203
|
METTLER TOLEDO™
30014096 |
InLab™ Expert Pro-ISM | Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology | BNC/RCA | Fixed Cable to sensor |
3725.00 DKK
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
||
Description
Ideal for critical samples such as suspensions or samples of unknown composition. Mettler Toledo Expert Series pH and pH/ATC Combination Electrodes with open junction puts the reference electrode in direct contact with the measured medium, eliminating the possibility of contamination or blockage.
- For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
- Features a PEEK plastic shaft for harsh environments and requires no maintenance
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- General Lab
- Beverages
- Waste water
- Suspended solids
- Environmental
- TRIS compatible
Accessories & Care
- Open Junction and non-refillable - no additional electrolyte required
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
PEEK | |
0°C to 100°C | |
ARGENTHAL | |
120 mm | |
12 mm |
0 to 14 | |
Xerolyt Polymer | |
U Glass | |
0.47 in. |