Learn More
HYDRANAL™-Moisture Test Kit, Honeywell Fluka™
Test kit for the visual water determination according to Karl Fischer without titrator
Marke: Honeywell Fluka 37858-1EA
Weitere Details : Weight : 0.10000kg Transport : UN number : 1993 Chem class : 3 Pack group : II
Beschreibung
Advantages of HYDRANAL- Auxiliaries
- Broad product range of solubilizer for special sample application
- Broad product range of buffer for pH related sample application
- Drying agents for cell protection and drying of carrier gasses.
Spezifikation
12°C (53.6°F) | |
1 Pc. | |
HYDRANAL™-Moisture Test Kit | |
UN1993BE |