Learn More
Brady™ Honeywell Xenon 1952G HD Bluetooth Scanner
The wireless area-imaging scanner Xenon 1952G HD delivers superior bar code scanning and digital image capture.
Brand:  Brady™ 306802
Additional Details : Weight : 0.15500kg Transport : UN number : 3481 Chem class : 9
Product Code. 17158654
  • 9050.00 DKK / Each
Estimated Shipment: 03-06-2024
to see stock.
Description
Description
  • Xenon 1952G HD incorporates a revolutionary decoding architecture and a custom sensor, enabling extended depth of field, faster reading, and improved scanning performance on poor quality bar codes
  • From high density linear to 2D bar codes found directly on the screen of a mobile device, Xenon 1952G HD decodes virtually all bar codes with ease
  • Xenon 1952G HD incorporates a Bluetooth Class 2, v2.1 radio, enabling unrestricted movement up to 10 m from the base, and a battery that provides up to 14 hours of battery life
  • For added convenience, a paging system helps locate misplaced scanners
Specifications
Specifications
High Density Imager Sensor
Lithium-ion Battery, 500 mA (at 5 V DC)
Bluetooth, USB