Learn More
Honeywell KCL™ Honeywell KCL™ 740-ermatril™ Gloves
Click to view available options
Size: S
Description
- AQL<0.65 free from natural latex resistant against a multitude of cytostatic drugs
- Virus-resistant according to ASTM 1671:2007
- Disposable protective glove excellent sense of touch
- Extremely comfortable
- Increase tear resistance
- Roughened fingertips for improved grip
Specifications
Certifications/Compliance | IFA 981069 |
Description | Single-use powder-free nitrile glove |
Color | Blue |
Disposable | Single-use |
Latex Free | Yes |
Material | Nitrile |
Powdered | Powder Free |
Size | Small |
Thickness (Metric) | 0.11 cm |
Glove Cuff Color | Blue |