Learn More
Thermo Scientific™ GC Liner Sealing Rings for Thermo Scientific™ and Agilent Instruments
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.
35.92 DKK - 886.94 DKK
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | For Use With (Equipment) | Price | Quantity & Availability | ||||
10561115
|
Thermo Scientific™
29033406 |
Liner Sealing Ring | TRACE and FOCUS GC SSL Injectors |
886.94 DKK
Pack of 10 |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
10467894
|
Thermo Scientific™
290GA243 |
Graphite Liner Sealing Ring | Agilent SSL Liners |
540.57 DKK
Pack of 10 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
||||
10638905
|
Thermo Scientific™
290OA241 |
Viton Liner Sealing Ring | TRACE 1300 Series GC and Agilent SSL Injectors |
194.80 DKK
Pack of 10 |
In Stock
Log in to see stock. |
||||
13415099
|
Thermo Scientific™
29001318 |
Liner Sealing Ring | TRACE 1300 Series GC PTV Injectors |
35.92 DKK
Each |
In Stock
Log in to see stock. |
||||
15398384
|
Thermo Scientific™
29001320 |
Liner Sealing Ring | TRACE 1300 Series GC SSL Injectors |
287.20 DKK
Pack of 5 |
In Stock
Log in to see stock. |
||||
10259314
|
Thermo Scientific™
290GA244 |
Graphite Liner Sealing Ring | Agilent 2.0 mm Liners |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10479234
|
Thermo Scientific™
290GA242 |
Graphite Liner Sealing Ring | Agilent Direct Straight Liners |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Match the best sealing ring with the injection port liner used in your Thermo Scientific™ or Agilent GC system.