Learn More
Thermo Scientific™ Fluid Transfer Assembly, Y Set
Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
Brand:  Thermo Scientific™ SH31104.01
Additional Details : Weight : 0.70000kg
Product Code. 15939856
  • POA / 1 unit
Estimated Shipment: 08-08-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process.
  • One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
  • Multiple manufacturing sites ensure security in product supply
  • Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
  • Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Specifications
3 x 12.7 mm (1/2 in.) ID x 19.05 mm (3/4 in.) wall; TPE 374; 2 x 0.51 m (20 in.) to 0.51 m (20 in.); MPX body-insert