Learn More
Fisherbrand™ Porcelain Crucible Covers
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles.
284.28 DKK - 1095.00 DKK
Specifications
Description | With Perforated handle |
---|---|
Material | Porcelain |
Finish | Glazed |
For Use With (Application) | Use with Fisherbrand high and wide form porcelain crucible |
Max. Temperature (Metric) | 1150°C |
Product Code | Brand | Height (Metric) | Thickness (Metric) | Diameter (Metric) Outer | Diameter (Metric) Inner | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Thickness (Metric) | Diameter (Metric) Outer | Diameter (Metric) Inner | Price | Quantity & Availability | ||
13439709
|
Fisherbrand™
|
5.5mm | 1.75mm | 40mm | 34mm |
296.00 DKK
Pack of 6 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||
13429709
|
Fisherbrand™
|
6mm | 1.9mm | 44mm | 38mm |
303.00 DKK
Pack of 6 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||
13449709
|
Fisherbrand™
|
7mm | 2.3mm | 53mm | 45mm |
308.00 DKK
Pack of 6 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||
13469709
|
Fisherbrand™
|
7mm | 2.5mm | 75mm | 66mm |
343.00 DKK
Pack of 6 |
Estimated Shipment: 20-09-2024
Log in to see stock. |
||
13459709
|
Fisherbrand™
|
7mm | 2.5mm | 60mm | 54mm |
1005.00 DKK
Pack of 6 |
Estimated Shipment: 10-07-2024
Log in to see stock. |
||
13479709
|
Fisherbrand™
|
8mm | 2.75mm | 87mm | 77mm |
1095.00 DKK
Pack of 6 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
||
13499709
|
Fisherbrand™
|
9mm | 3mm | 115mm | 105mm |
506.00 DKK
Pack of 6 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||
13419709
|
Fisherbrand™
|
5mm | 1.75mm | 30mm | 26mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
13489709
|
Fisherbrand™
|
8mm | 2.75mm | 99mm | 90mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
For use with Fisherbrand High- and Wide-Form Porcelain Crucibles
- Glazed inside and out except for the outside bottom surface
- With perforated handle
- Strongly recommended that heating and cooling rates not exceed 200°C per hour and maximum use temperature of 1100°C
Specifications
With Perforated handle | |
Glazed | |
1150°C |
Porcelain | |
Use with Fisherbrand high and wide form porcelain crucible | |
Crucible Cover |