Learn More
Fisherbrand™ Elite™ Adjustable-Volume Pipettes
Extremely low plunger forces work longer without fatigue and reduce the risk of RSI.
1400.00 DKK - 2160.00 DKK
Specifications
Autoclavable | Autoclavable |
---|---|
Product Line | Elite™ |
Finger Style | Ergonomic |
Includes | Calibration certificate, in-lab calibration tool |
Secure Tip Attachment | Soft touch tip ejection |
Includes: Calibration certificate and in-lab calibration tool.
Product Code | Brand | Accuracy | Volume (Metric) | Increments | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Accuracy | Volume (Metric) | Increments | Price | Quantity & Availability | |||
11815762
|
Fisherbrand™
|
2.5 to 12% | 0.2 to 2 μL | 0.002μL |
1880.00 DKK
Each |
In Stock
Log in to see stock. |
|||
11825762
|
Fisherbrand™
|
1.5 to 6% | 0.5 to 5 μL | 0.01μL |
1400.00 DKK
Each |
In Stock
Log in to see stock. |
|||
11835762
|
Fisherbrand™
|
1 to 2.5% | 1 to 10 μL | 0.02μL |
1400.00 DKK
Each |
In Stock
Log in to see stock. |
|||
11845762
|
Fisherbrand™
|
1 to 3% | 2 to 20 μL | 0.02μL |
2095.00 DKK
Each |
In Stock
Log in to see stock. |
|||
11855762
|
Fisherbrand™
|
0.6 to 3% | 5 to 50 μL | 0.1μL |
1935.00 DKK
Each |
In Stock
Log in to see stock. |
|||
11865762
|
Fisherbrand™
|
0.8 to 3% | 10 to 100 μL | 0.2μL |
2095.00 DKK
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
|||
11875762
|
Fisherbrand™
|
0.6 to 1.8% | 20 to 200 μL | 0.2μL |
2095.00 DKK
Each |
In Stock
Log in to see stock. |
|||
11805772
|
Fisherbrand™
|
0.5 to 2% | 1 to 10 mL | 0.02mL |
2095.00 DKK
Each |
In Stock
Log in to see stock. |
|||
11895762
|
Fisherbrand™
|
0.5 to 2% | 0.5 to 5 mL | 0.01mL |
2095.00 DKK
Each |
In Stock
Log in to see stock. |
|||
11885762
|
Fisherbrand™
|
0.6 to 1% | 100 to 1000 μL | 1μL |
2160.00 DKK
Each |
In Stock
Log in to see stock. |
|||
Description
Extremely low plunger forces work longer without fatigue and reduce the risk of RSI.
- Fully autoclavable — entire pipette can be steam autoclaved
- Soft-touch tip ejection — a gearing mechanism reducing tip ejection force up to 50%
- Ergonomic comfortable lightweight handle — comfortable grip and effortless tip attachment
- Optimal fit with SureOne™ pipette tips — best pipetting performance
- Super blow-out piston — creates 150% bigger air push than conventional piston for delivery of the micro size drops
Warranty
Three years.
Specifications
Autoclavable | |
Ergonomic | |
Soft touch tip ejection |
Elite™ | |
Calibration certificate, in-lab calibration tool | |
3 Years |