Learn More
Lenz Laborglasinstrumente™ DURAN™ Glass Round Bottom Flasks
Can be used to make solutions or hold, contain, collect and measure chemicals. Lenz Laborglasinstrumente™ DURAN™ Glass Round Bottom Flasks are manufactured from durable borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ B0418021
Additional Details : Weight : 0.01800kg
Product Code. 10224460
  • 91.00 DKK / Each
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
Description
Description
  • With inscription label
  • Available in a range of capacities
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
10 mL
Borosilicate Glass, Duran Glass
Head Touches NSH 19/26
Round