Learn More
Invitrogen™ Dual LED Blue/White Light Transilluminator
Innovatively designed dual-light source transilluminator for visualization of common nucleic acid stains, colorimetric protein and membrane stains, and SYPRO stains.
Brand: Invitrogen™ LB0100
5591.16 DKK valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Additional Details : Weight : 1.81000kg
Description
This Dual LED Blue/White Light Transilluminator is an innovatively designed dual-light source transilluminator. It is designed for visualization of nucleic acid gels, protein gels, and membrane stains.
Dual LED Blue/White Light Transilluminator features include:
• White and blue light sources that allow for visualization of common nucleic acid stains, colorimetric protein and membrane stains, and SYPRO protein stains
• Magnetic filter screen with hinge-free design for simplicity and safety, allowing observation and gel-cutting without protective goggles
• Five-minute automatic power-off and long-lasting LEDs that are safer for the eyes and skin compared with UV bulb illumination
• Three-level adjustable light intensity to achieve the best observational or imaging quality
• Compact design for ease of mobility and storage
Specifications
LED Blue and White | |
Blue LED Emission 470 nm, Amber Filter at 580 nm | |
Transilluminator | |
1.814 kg |
3-Levels of Adjustment | |
UL/ETL | |
18.5 x 22 x 3.0 cm; 7.28 x 8.66 x 1.18 in | |
100/240 V |