Learn More
Bochem™ Dissecting Set
Stainless Steel. Bochem™ Dissecting Set includes eight pieces.
Brand:  Bochem™ 12017
Additional Details : Weight : 0.50000kg
Product Code. 10430733
  • 325.00 DKK / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
Description
Description
  • Stainless Steel
  • Eight pieces
Specifications
Specifications
Scissors, Blunt Forceps, Scapel with blade (35mm), Scapel with blade (45mm), Dressing scissors, Sharp Forceps, Dissecting Needle, Lancet